Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lancl3
Entrez Gene ID
236285
Gene Name
LanC lantibiotic synthetase component C-like 3 (bacterial)
Gene Aliases
6030463G20Rik
Chromosome Location
Chr.X: 9199973 - 9268085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173414.3 NP_775590.2 3 - 4 1157 104
GenBank mRNA AK031623.1 - 3 - 4 1158 104
AK043841.1 - 3 - 4 358 104
AK045254.1 - 3 - 4 745 104
BC115944.1 - 2 - 3 280 104

View Full Product Details