The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173418.3 | NP_775594.1 | 2 - 3 | 918 | 103 |
GenBank mRNA | AK044383.1 | - | 2 - 3 | 919 | 103 |
AK044611.1 | - | 2 - 3 | 920 | 103 | |
BC037728.1 | - | 2 - 3 | 818 | 103 | |
BC057628.1 | - | 2 - 3 | 893 | 103 |