The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173433.2 | NP_775609.2 | 1 - 2 | 371 | 90 |
GenBank mRNA | AK030020.1 | - | 1 - 2 | 371 | 90 |