Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ephb1
Entrez Gene ID
270190
Gene Name
Eph receptor B1
Gene Aliases
9330129L11, AW488255, C130099E04Rik, Cek6, ENSMUSG00000074119, Elk, Elkh, Hek6, Net
Chromosome Location
Chr.9: 101922128 - 102354693 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168296.1 NP_001161768.1 13 - 14 2925 61
NM_173447.3 NP_775623.3 14 - 15 3048 61
GenBank mRNA AK033966.1 - 14 - 15 3048 61
AK036148.1 - 14 - 15 2736 61
AK036211.1 - 13 - 14 2924 61
AK135018.1 - 14 - 15 3014 61
BC057301.1 - 14 - 15 2988 61

View Full Product Details