The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175309.4 | NP_780518.1 | 5 - 6 | 982 | 89 |
GenBank mRNA | AK042067.1 | - | 5 - 6 | 778 | 89 |
AK052525.1 | - | 5 - 6 | 984 | 89 | |
AK157647.1 | - | 5 - 6 | 789 | 89 | |
AY233463.1 | - | 5 - 6 | 776 | 89 | |
BC120594.1 | - | 5 - 6 | 732 | 89 | |
BC137742.1 | - | 5 - 6 | 732 | 89 |