Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdca2
Entrez Gene ID
108912
Gene Name
cell division cycle associated 2
Gene Aliases
2610311M19Rik, AI586158
Chromosome Location
Chr.14: 67673180 - 67715841 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110162.1 NP_001103632.1 1 - 2 388 88
GenBank mRNA AK031083.1 - 1 - 2 154 88
AK160223.1 - 1 - 2 150 88
AK166952.1 - 1 - 2 157 88
BC037136.1 - 1 - 2 188 88
BC067009.1 - 1 - 2 119 88
CD354922.1 - 1 - 2 388 88

View Full Product Details