Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptgds
Entrez Gene ID
19215
Gene Name
prostaglandin D2 synthase (brain)
Gene Aliases
21kDa, L-PGDS, PGD2, PGDS, PGDS2, Ptgs3
Chromosome Location
Chr.2: 25466709 - 25470109 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008963.2 NP_032989.2 1 - 2 191 57
GenBank mRNA AK131859.1 - 1 - 2 191 57
BC043015.1 - 1 - 2 177 57
X89222.1 - 1 - 2 162 57

View Full Product Details