Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp7b
Entrez Gene ID
11979
Gene Name
ATPase, Cu++ transporting, beta polypeptide
Gene Aliases
Atp7a, WND, tx
Chromosome Location
Chr.8: 21992783 - 22060074 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007511.2 NP_031537.2 9 - 10 2471 90

View Full Product Details