The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163478.1 | NP_001156950.1 | 7 - 8 | 741 | 98 |
NM_001163479.1 | NP_001156951.1 | 5 - 6 | 679 | 98 | |
NM_011231.2 | NP_035361.2 | 6 - 7 | 879 | 98 | |
NR_028094.1 | 8 - 9 | 823 | 98 | ||
NR_028095.1 | 7 - 8 | 961 | 98 | ||
NR_028096.1 | 7 - 8 | 977 | 98 | ||
NR_028097.1 | 6 - 7 | 761 | 98 | ||
NR_028098.1 | 5 - 6 | 663 | 98 | ||
GenBank mRNA | AF231926.1 | - | 7 - 8 | 817 | 98 |
AK010690.1 | - | 5 - 6 | 431 | 98 | |
AK160162.1 | - | 7 - 8 | 728 | 98 | |
BC003948.1 | - | 7 - 8 | 804 | 98 | |
BC021335.1 | - | 6 - 7 | 725 | 98 | |
BC057661.1 | - | 6 - 7 | 1393 | 98 | |
BC132473.1 | - | 6 - 7 | 757 | 98 | |
BC138547.1 | - | 6 - 7 | 757 | 98 | |
BC145323.1 | - | 7 - 8 | 839 | 98 | |
BC145324.1 | - | 6 - 7 | 741 | 98 | |
BC145325.1 | - | 7 - 8 | 855 | 98 | |
BC145326.1 | - | 7 - 8 | 815 | 98 | |
CN456589.1 | - | 5 - 6 | 588 | 98 | |
CV562676.1 | - | 6 - 7 | 607 | 98 | |
U12922.1 | - | 6 - 7 | 735 | 98 |