Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dis3l2
Entrez Gene ID
208718
Gene Name
DIS3 mitotic control homolog (S. cerevisiae)-like 2
Gene Aliases
4930429A22Rik, 8030493P09Rik
Chromosome Location
Chr.1: 86703779 - 87050097 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172157.1 NP_001165628.1 21 - 22 2696 67
NM_153530.2 NP_705758.1 20 - 21 2654 67
GenBank mRNA AK031180.1 - 21 - 22 2679 67
BC036177.1 - 20 - 21 2615 67

View Full Product Details