The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153749.4 | NP_715630.3 | 3 - 4 | 705 | 71 |
GenBank mRNA | AB086265.1 | - | 3 - 4 | 470 | 71 |
AK029010.1 | - | 3 - 4 | 706 | 71 | |
AK030695.1 | - | 3 - 4 | 629 | 71 | |
BC110547.1 | - | 3 - 4 | 681 | 71 | |
BC110548.1 | - | 3 - 4 | 681 | 71 |