Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thap3
Entrez Gene ID
69876
Gene Name
THAP domain containing, apoptosis associated protein 3
Gene Aliases
2010013E08Rik, 2210418H06Rik
Chromosome Location
Chr.4: 151982638 - 151988986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175152.4 NP_780361.2 2 - 3 294 91
GenBank mRNA AK008225.1 - 2 - 3 266 91
AK038184.1 - 2 - 3 268 91
AK050740.1 - 2 - 3 308 91
BC094395.1 - 2 - 3 275 91
BC115485.1 - 2 - 3 292 91
BC115486.1 - 2 - 3 292 91
BY013572.1 - 2 - 3 294 91

View Full Product Details