The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038993.3 | NP_001034082.1 | 9 - 10 | 1290 | 85 |
NM_175201.5 | NP_780410.2 | 8 - 9 | 1320 | 85 | |
GenBank mRNA | AK147608.1 | - | 9 - 10 | 1347 | 85 |
AK163346.1 | - | 4 - 5 | 451 | 85 | |
BC060730.1 | - | 8 - 9 | 1297 | 85 | |
BC062976.1 | - | 8 - 9 | 1234 | 85 |