Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Papl
Entrez Gene ID
101744
Gene Name
iron/zinc purple acid phospatase-like
Gene Aliases
AU017076, C330005M16Rik
Chromosome Location
Chr.7: 28607634 - 28631361 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175319.4 NP_780528.1 1 - 2 172 78
GenBank mRNA AK049131.1 - 1 - 2 173 78
BC094908.1 - 1 - 2 68 78
BC132375.1 - 1 - 2 159 78
BC138307.1 - 1 - 2 158 78

View Full Product Details