The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175333.3 | NP_780542.1 | 4 - 5 | 495 | 106 |
GenBank mRNA | AK077111.1 | - | 4 - 5 | 496 | 106 |
BC115588.1 | - | 5 - 6 | 594 | 106 | |
BC115589.1 | - | 4 - 5 | 488 | 106 | |
FM165286.1 | - | 4 - 5 | 437 | 106 |