The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175374.3 | NP_780583.1 | 1 - 2 | 422 | 69 |
GenBank mRNA | AK036424.1 | - | 1 - 2 | 326 | 69 |
AK078897.1 | - | 1 - 2 | 390 | 69 |