The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175548.3 | NP_780757.1 | 4 - 5 | 690 | 71 |
GenBank mRNA | AK030681.1 | - | 4 - 5 | 996 | 71 |
AK044845.1 | - | 4 - 5 | 690 | 71 | |
AK140043.1 | - | 4 - 5 | 827 | 71 | |
AK158702.1 | - | 4 - 5 | 1174 | 71 | |
AK164171.1 | - | - | 997 | 71 |