The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175678.3 | NP_783609.1 | 1 - 2 | 50 | 61 |
GenBank mRNA | AK033957.1 | - | 1 - 2 | 51 | 61 |