Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhbdd3
Entrez Gene ID
279766
Gene Name
rhomboid domain containing 3
Gene Aliases
Ptag
Chromosome Location
Chr.11: 5095631 - 5106100 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290491.1 NP_001277420.1 3 - 4 637 66
NM_001290492.1 NP_001277421.1 3 - 4 649 66
NM_001290493.1 NP_001277422.1 2 - 3 254 66
NM_177370.4 NP_796344.1 3 - 4 637 66
NR_110961.1 2 - 3 343 66
GenBank mRNA AK053223.1 - 3 - 4 472 66
AK077457.1 - 3 - 4 473 66
AK156451.1 - 3 - 4 491 66
BC062155.1 - 1 - 2 352 66
BC096693.1 - 2 - 3 263 66
BC132459.1 - 2 - 3 302 66
BC138479.1 - 2 - 3 302 66
BC145287.1 - 2 - 3 314 66
BC145289.1 - 2 - 3 302 66
BC145290.1 - 2 - 3 302 66

View Full Product Details