The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177585.3 | NP_808253.1 | 3 - 4 | 250 | 70 |
GenBank mRNA | AK052136.1 | - | 3 - 4 | 251 | 70 |
BC107305.1 | - | 3 - 4 | 214 | 70 | |
BC107306.1 | - | 3 - 4 | 214 | 70 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113419.2 | NP_001106890.1 | 3 - 4 | 250 | 70 |
GenBank mRNA | KJ941154.1 | - | 3 - 4 | 238 | 70 |