Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdzd7
Entrez Gene ID
100503041
Gene Name
PDZ domain containing 7
Gene Aliases
9130207N01, EG435601, Pdzk7
Chromosome Location
Chr.19: 45026907 - 45058308 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195265.1 NP_001182194.1 14 - 15 2732 78
GenBank mRNA AK033653.1 - 4 - 5 627 78
BC096488.1 - 4 - 5 848 78
BC147398.1 - 1 - 2 535 78
BC147399.1 - 1 - 2 535 78
BC172054.1 - 1 - 2 535 78
KF041446.1 - 15 - 16 2698 78
KF041447.1 - 15 - 16 2727 78
KF041448.1 - 12 - 13 2512 78
KF041449.1 - 13 - 14 2545 78
KF041450.1 - 14 - 15 2637 78
KF041451.1 - 15 - 16 2518 78

View Full Product Details