Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ric1
Entrez Gene ID
226089
Gene Name
RAB6A GEF complex partner 1
Gene Aliases
C030046E11Rik, C130057E09Rik, Kiaa1432
Chromosome Location
Chr.19: 29522282 - 29605921 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081319.1 NP_001074788.1 3 - 4 571 135
GenBank mRNA AK048394.1 - 3 - 4 519 135
AK156653.1 - 3 - 4 472 135
AK173171.1 - 2 - 3 153 135
BC125448.1 - 3 - 4 468 135
BC132346.1 - 3 - 4 468 135

View Full Product Details