The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178378.3 | NP_848465.1 | 8 - 9 | 995 | 101 |
GenBank mRNA | AK010269.1 | - | 5 - 6 | 544 | 101 |
AK146422.1 | - | 6 - 7 | 733 | 101 | |
BC049990.1 | - | 8 - 9 | 979 | 101 |