Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wipi2
Entrez Gene ID
74781
Gene Name
WD repeat domain, phosphoinositide interacting 2
Gene Aliases
1110018O08Rik, 2510001I10Rik
Chromosome Location
Chr.5: 142629554 - 142669672 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178398.4 NP_848485.1 7 - 8 867 99
GenBank mRNA AK010881.1 - 7 - 8 893 99
AK155113.1 - 7 - 8 897 99
AK169051.1 - 7 - 8 841 99
AK172030.1 - 7 - 8 843 99
BC044894.1 - 7 - 8 892 99

View Full Product Details