Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlrx1
Entrez Gene ID
270151
Gene Name
NLR family member X1
Gene Aliases
BC034204, NOD9
Chromosome Location
Chr.9: 44252713 - 44268599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163742.1 NP_001157214.1 9 - 10 2758 85
NM_001163743.1 NP_001157215.1 9 - 10 2859 85
NM_178420.3 NP_848507.2 9 - 10 2847 85
GenBank mRNA AK141296.1 - 9 - 10 2831 85
AK145988.1 - 9 - 10 2848 85
AK166689.1 - 9 - 10 2685 85
AK170088.1 - 9 - 10 2803 85
BC034204.1 - 8 - 9 2641 85
BC050054.1 - 9 - 10 2754 85

View Full Product Details