Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbms3
Entrez Gene ID
207181
Gene Name
RNA binding motif, single stranded interacting protein
Gene Aliases
6720477E09Rik, 8430436O14Rik
Chromosome Location
Chr.9: 116572746 - 118040292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172121.1 NP_001165592.1 12 - 13 1233 91
NM_001172123.1 NP_001165594.1 12 - 13 1756 91
NM_001172124.1 NP_001165595.1 12 - 13 1756 91
NM_178660.4 NP_848775.2 11 - 12 1705 91
GenBank mRNA AK030478.1 - 11 - 12 1365 91
AK032938.1 - 11 - 12 1702 91
AK033005.1 - 12 - 13 1399 91
AK046197.1 - 12 - 13 1234 91
AK052196.1 - 12 - 13 1737 91
BC108402.1 - 11 - 12 1679 91
BC117841.1 - 11 - 12 1145 91

View Full Product Details