The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178664.5 | NP_848779.2 | 2 - 3 | 283 | 89 |
GenBank mRNA | AK031361.1 | - | 2 - 3 | 272 | 89 |
AK043722.1 | - | 2 - 3 | 286 | 89 | |
AK045461.1 | - | 2 - 3 | 205 | 89 | |
AK156318.1 | - | 2 - 3 | 261 | 89 | |
BB641432.1 | - | 2 - 3 | 283 | 89 | |
BC088993.1 | - | 2 - 3 | 169 | 89 | |
BC125626.1 | - | 2 - 3 | 256 | 89 | |
BC132193.1 | - | 2 - 3 | 256 | 89 | |
BC145163.1 | - | 2 - 3 | 260 | 89 | |
BC145165.1 | - | 2 - 3 | 256 | 89 | |
BC145166.1 | - | 2 - 3 | 256 | 89 |