The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178678.4 | NP_848793.1 | 2 - 3 | 2080 | 75 |
GenBank mRNA | AK032485.1 | - | 2 - 3 | 2097 | 75 |
AK035773.1 | - | 2 - 3 | 2077 | 75 | |
AK035858.1 | - | 2 - 3 | 2083 | 75 | |
AK035882.1 | - | 2 - 3 | 2081 | 75 | |
AK036180.1 | - | 2 - 3 | 2100 | 75 | |
AK163753.1 | - | 2 - 3 | 2073 | 75 | |
BC113178.1 | - | 2 - 3 | 1754 | 75 | |
BC113784.1 | - | 2 - 3 | 1754 | 75 |