The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178708.3 | NP_848823.2 | 2 - 3 | 663 | 123 |
GenBank mRNA | AK042945.1 | - | 2 - 3 | 153 | 123 |
AK045796.1 | - | 2 - 3 | 143 | 123 | |
AK048722.1 | - | 2 - 3 | 154 | 123 | |
AK082891.1 | - | 2 - 3 | 729 | 124 | |
AK163697.1 | - | 2 - 3 | 150 | 123 | |
BC006047.1 | - | 2 - 3 | 152 | 123 | |
BC118533.1 | - | 2 - 3 | 663 | 123 |