The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159963.1 | NP_001153435.1 | 6 - 7 | 998 | 132 |
NM_178729.4 | NP_848844.1 | 6 - 7 | 998 | 132 | |
GenBank mRNA | AK029957.1 | - | 6 - 7 | 946 | 132 |
AK041497.1 | - | 6 - 7 | 970 | 132 | |
AK082931.1 | - | 6 - 7 | 968 | 132 | |
AK085100.1 | - | 6 - 7 | 971 | 132 | |
AK088067.1 | - | 6 - 7 | 965 | 132 | |
AK159744.1 | - | 6 - 7 | 987 | 132 | |
AK171783.1 | - | 6 - 7 | 858 | 132 | |
BC023291.1 | - | 4 - 5 | 603 | 132 | |
BC047214.1 | - | - | 911 | 132 | |
BC113798.1 | - | 6 - 7 | 916 | 132 |