Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc8a1
Entrez Gene ID
20541
Gene Name
solute carrier family 8 (sodium/calcium exchanger), member 1
Gene Aliases
AI852629, AV344025, D930008O12Rik, Ncx1
Chromosome Location
Chr.17: 81373105 - 81738387 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112798.2 NP_001106269.1 9 - 9 17389 68
NM_001286684.1 NP_001273613.1 12 - 12 14663 68
NM_011406.3 NP_035536.2 11 - 11 17425 68
NR_104580.1 9 - 9 17424 68
GenBank mRNA AK046123.1 - 1 - 1 66 68
AK078052.1 - 1 - 1 2456 68
BC060264.1 - 1 - 1 1406 68
BC094585.1 - 1 - 1 1640 68
BC096032.1 - 2 - 2 1254 68

View Full Product Details