Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmat2
Entrez Gene ID
66492
Gene Name
zinc finger, matrin type 2
Gene Aliases
2610510D14Rik, 2900082I05Rik
Chromosome Location
Chr.18: 36793923 - 36799660 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025594.3 NP_079870.1 2 - 3 134 76
GenBank mRNA AK008717.1 - 2 - 3 127 76
BC024881.1 - 2 - 3 134 76
BC089501.1 - 2 - 3 129 76

View Full Product Details