Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpatch2
Entrez Gene ID
67769
Gene Name
G patch domain containing 2
Gene Aliases
5830433G22Rik, 5830436K05Rik, AI427714, AI447508, AW107440, AW491060, Gpatc2
Chromosome Location
Chr.1: 187215393 - 187369480 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026367.4 NP_080643.4 2 - 3 885 85
GenBank mRNA AK017975.1 - 2 - 3 854 85
AK029990.1 - 2 - 3 869 85
AK030026.1 - 2 - 3 884 85
AK053781.1 - 2 - 3 877 85
AK167826.1 - 2 - 3 889 85
AK169660.1 - 2 - 3 886 85
BC054810.1 - 2 - 3 857 85

View Full Product Details