Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nwd1
Entrez Gene ID
319555
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
A230063L24Rik
Chromosome Location
Chr.8: 72646225 - 72716880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176940.5 NP_795914.3 12 - 13 3242 93
GenBank mRNA AK034868.1 - - 2941 93
AK038792.1 - 11 - 12 2935 93
BC082552.1 - 12 - 13 3182 93
BC144786.1 - 10 - 11 2832 93
BC144787.1 - 10 - 11 2832 93
BC145703.1 - 10 - 11 2832 93

View Full Product Details