The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177005.4 | NP_795979.2 | 3 - 4 | 347 | 131 |
GenBank mRNA | AK050379.1 | - | 3 - 4 | 348 | 131 |
BC115464.1 | - | 3 - 4 | 346 | 131 |