Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Glt1d1
- Entrez Gene ID
-
319804
- Gene Name
- glycosyltransferase 1 domain containing 1
- Gene Aliases
-
5730455A04Rik
- Chromosome Location
-
Chr.5: 127632130 - 127708750 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK050379.1 |
- |
6 - 7 |
553 |
68 |
View Full Product Details