The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166685.1 | NP_001160157.1 | 7 - 8 | 879 | 115 |
NM_177088.3 | NP_796062.2 | 8 - 9 | 1008 | 115 | |
GenBank mRNA | AK052705.1 | - | 8 - 9 | 936 | 115 |
AK076400.1 | - | 8 - 9 | 986 | 115 | |
BC090675.1 | - | 8 - 9 | 964 | 115 | |
BC116308.1 | - | 7 - 8 | 845 | 115 | |
BC116309.1 | - | 7 - 8 | 858 | 115 |