The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177448.4 | NP_803231.1 | 1 - 2 | 139 | 62 |
GenBank mRNA | AK049560.1 | - | 1 - 2 | 102 | 62 |
AK133854.1 | - | 1 - 2 | 147 | 62 | |
AY157609.1 | - | 1 - 2 | 96 | 62 | |
BC052831.1 | - | 1 - 2 | 143 | 62 |