The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177677.3 | NP_808345.1 | 3 - 4 | 428 | 83 |
GenBank mRNA | AK076638.1 | - | 3 - 4 | 429 | 83 |
BC089502.1 | - | 3 - 4 | 407 | 83 |