The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271879.1 | NP_001258808.1 | 15 - 16 | 1833 | 90 |
NM_177694.6 | NP_808362.2 | 16 - 17 | 1983 | 90 | |
NR_073508.1 | 16 - 17 | 1976 | 90 | ||
GenBank mRNA | AB125740.1 | - | 16 - 17 | 1983 | 90 |
AB206762.1 | - | 16 - 17 | 1983 | 90 | |
AK034197.1 | - | 16 - 17 | 1965 | 90 | |
BC109163.2 | - | 15 - 16 | 1755 | 90 |