Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lsm14b
Entrez Gene ID
241846
Gene Name
LSM14 homolog B (SCD6, S. cerevisiae)
Gene Aliases
9430078B04, BC040823
Chromosome Location
Chr.2: 180024902 - 180035466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177727.4 NP_808395.2 4 - 5 793 77
GenBank mRNA AK035040.1 - 4 - 5 822 77
BC040823.1 - 4 - 5 793 77
CF730033.1 - 4 - 5 524 77

View Full Product Details