Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubash3a
Entrez Gene ID
328795
Gene Name
ubiquitin associated and SH3 domain containing, A
Gene Aliases
5830413C03Rik, C330001M22, Sts-2, TULA, TULA-1
Chromosome Location
Chr.17: 31207713 - 31253419 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177823.4 NP_808491.2 9 - 10 1334 80
GenBank mRNA AK040572.1 - 9 - 10 1295 80
AK041690.1 - 9 - 10 1301 80
AK049106.1 - 8 - 9 1234 80
AK169696.1 - 9 - 10 1289 80

View Full Product Details