Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stk32a
Entrez Gene ID
269019
Gene Name
serine/threonine kinase 32A
Gene Aliases
A930015B13Rik, YANK1
Chromosome Location
Chr.18: 43207697 - 43317481 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178749.3 NP_848864.1 10 - 11 1145 134
GenBank mRNA AK036266.1 - 10 - 11 1146 134
AK042599.1 - 10 - 11 1259 134
AK044474.1 - 10 - 11 1155 134
BC055002.1 - - 1075 134

View Full Product Details