Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsm5
Entrez Gene ID
272428
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
C730019D22, C730027J19Rik
Chromosome Location
Chr.7: 119526131 - 119543360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178758.3 NP_848873.1 12 - 13 1726 109
GenBank mRNA AK050132.1 - 13 - 14 1823 109
AK050219.1 - 12 - 13 1751 109
AK050288.1 - 12 - 13 1746 109
AK050404.1 - 12 - 13 1786 109
AK050458.1 - 12 - 13 1688 109
AK149591.1 - 12 - 13 1746 109
BC095985.1 - 12 - 13 1726 109

View Full Product Details