Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acsm5
Entrez Gene ID
272428
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
C730019D22, C730027J19Rik
Chromosome Location
Chr.7: 119526131 - 119543360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178758.3 NP_848873.1 5 - 6 955 88
GenBank mRNA AK040650.1 - 5 - 6 892 88
AK050132.1 - 6 - 7 1052 88
AK050219.1 - 5 - 6 980 88
AK050288.1 - 5 - 6 88
AK050458.1 - 5 - 6 917 88
AK080219.1 - 5 - 6 891 88
AK149591.1 - 5 - 6 88
BC095985.1 - 5 - 6 955 88

View Full Product Details