The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285840.1 | NP_001272769.1 | 2 - 3 | 583 | 76 |
NM_176998.4 | NP_795972.2 | 3 - 4 | 784 | 76 | |
NM_178765.3 | NP_848880.2 | 3 - 4 | 681 | 76 | |
GenBank mRNA | AB232447.1 | - | 3 - 4 | 784 | 76 |
AB232449.1 | - | 3 - 4 | 681 | 76 | |
AK029072.1 | - | 2 - 3 | 280 | 76 | |
AK030482.1 | - | 3 - 4 | 413 | 76 | |
AK030720.1 | - | 3 - 4 | 413 | 76 | |
AK163432.1 | - | 3 - 4 | 446 | 76 | |
AK220268.1 | - | 2 - 3 | 583 | 76 | |
BC065045.1 | - | 3 - 4 | 678 | 76 |