Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cldn17
Entrez Gene ID
239931
Gene Name
claudin 17
Gene Aliases
-
Chromosome Location
Chr.16: 88505807 - 88506978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181490.3 NP_852467.1 1 - 1 651 67
GenBank mRNA AK048287.1 - 1 - 1 653 67

View Full Product Details