Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpbg
Entrez Gene ID
21983
Gene Name
trophoblast glycoprotein
Gene Aliases
5T4, AW495680, WAIF1
Chromosome Location
Chr.9: 85842380 - 85847055 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011627.4 NP_035757.2 1 - 2 214 66
GenBank mRNA AK051162.1 - 1 - 2 185 66
AK165597.1 - 1 - 2 218 66

View Full Product Details