The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022724.4 | NP_073561.2 | 1 - 2 | 288 | 131 |
GenBank mRNA | AF149205.1 | - | 1 - 2 | 266 | 131 |
AK083457.1 | - | 1 - 2 | 266 | 131 | |
BC032960.1 | - | 1 - 2 | 271 | 131 | |
CX227854.1 | - | 1 - 2 | 288 | 131 |