The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130866.4 | NP_570936.1 | 1 - 2 | 157 | 84 |
GenBank mRNA | AK014616.1 | - | 1 - 2 | 82 | 84 |
AK028467.1 | - | 1 - 2 | 82 | 84 |